- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:PEDF
- Description:Mouse PEDF ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Mouse PEDF ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse PEDF. No significant cross-reactivity or interference between PEDF and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.156 - 10 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.094 ng/ml
- Regulatory Status:RUO
- Cat. No.:76746-858
Mouse PEDF ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse PEDF in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 0.156 to 10 ng/ml
- Sensitivity: 0.094 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Mouse PEDF ELISA Kit (A77126) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse PEDF in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PEDF has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PEDF present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PEDF Antibody, which binds the captured PEDF present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PEDF captured in each well. The concentration of PEDF can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.