- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Goat
- Target protein:Superoxide Dismutase 1
- Description:Goat superoxide dismutase 1 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross Reactivity:Goat Superoxide Dismutase 1 ELISA Kit exhibits high specificity and excellent specificity for the detection of goat Superoxide Dismutase 1. No significant cross-reactivity or interference between Superoxide Dismutase 1 and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.781 - 50 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.469 ng/ml
- Regulatory Status:RUO
- Cat. No.:76746-808
Goat Superoxide Dismutase 1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of goat Superoxide Dismutase 1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Goat Superoxide Dismutase 1 ELISA kit (A247354) employs the sandwich enzyme immunoassay technique for the quantitative measurement of goat Superoxide Dismutase 1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Superoxide Dismutase 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Superoxide Dismutase 1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Superoxide Dismutase 1 Antibody, which binds the captured Superoxide Dismutase 1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Superoxide Dismutase 1 captured in each well. The concentration of Superoxide Dismutase 1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.