Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Stabilin 2 ELISA Kit
Rat Stabilin 2 ELISA Kit
Catalog # 76743-958
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Rat Stabilin 2 ELISA Kit
Catalog # 76743-958
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A79725-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Stabilin 2
  • Description:
    Rat stabilin 2 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Rat Stabilin 2 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Stabilin 2. No significant cross-reactivity or interference between Stabilin 2 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.313 - 20 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.188 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76743-958

Specifications

About this item

Rat Stabilin 2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Stabilin 2 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 0.313 to 20 ng/ml
  • Sensitivity: 0.188 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat Stabilin 2 ELISA kit (A79725) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Stabilin 2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Stabilin 2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Stabilin 2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Stabilin 2 Antibody, which binds the captured Stabilin 2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Stabilin 2 captured in each well. The concentration of Stabilin 2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.