Order Entry
Puerto Rico
ContactUsLinkComponent
Human Calpain Small Subunit 1 ELISA Kit
Human Calpain Small Subunit 1 ELISA Kit
Catalog # 76742-208
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human Calpain Small Subunit 1 ELISA Kit
Catalog # 76742-208
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A87008-96
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Calpain Small Subunit 1
  • Description:
    Human Calpain 4 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human Calpain Small Subunit 1 ELISA Kit exhibits high specificity and excellent specificity for the detection of human Calpain Small Subunit 1. No significant cross-reactivity or interference between Calpain Small Subunit 1 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.094 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76742-208

Specifications

About this item

Human Calpain Small Subunit 1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Calpain Small Subunit 1 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 0.156 to 10 ng/ml
  • Sensitivity: 0.094 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human Calpain Small Subunit 1 ELISA Kit (A87008) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Calpain Small Subunit 1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Calpain Small Subunit 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Calpain Small Subunit 1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Calpain Small Subunit 1 Antibody, which binds the captured Calpain Small Subunit 1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Calpain Small Subunit 1 captured in each well. The concentration of Calpain Small Subunit 1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.