- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:PKA beta (catalytic subunit)
- Description:Mouse PKA beta (catalytic subunit) ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Mouse PKA beta (catalytic subunit) ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse PKA beta (catalytic subunit). No significant cross-reactivity or interference between PKA beta (catalytic subunit) and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.156 - 10 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.094 ng/ml
- Regulatory Status:RUO
- Cat. No.:76741-308
Mouse PKA beta (catalytic subunit) ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse PKA beta (catalytic subunit) in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 0.156 to 10 ng/ml
- Sensitivity: 0.094 ng/ml
- Sample type: Serum. plasma. tissue homogenates. and other biological fluids
- Assay precision: Intra assay: CV <8%. Inter assay: CV <10%
Mouse PKA beta (catalytic subunit) ELISA Kit (A75754) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse PKA beta (catalytic subunit) in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PKA beta (catalytic subunit) has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PKA beta (catalytic subunit) present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PKA beta (catalytic subunit) Antibody, which binds the captured PKA beta (catalytic subunit) present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PKA beta (catalytic subunit) captured in each well. The concentration of PKA beta (catalytic subunit) can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.