- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Prostaglandin E Synthase/MPGES-1
- Description:Mouse prostaglandin E synthase/MPGES-1 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Mouse Prostaglandin E Synthase/MPGES-1 ELISA kit exhibits high specificity and excellent specificity for the detection of mouse Prostaglandin E Synthase/MPGES-1. No significant cross-reactivity or interference between Prostaglandin E Synthase/MPGES-1 and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.156 - 10 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.094 ng/ml
- Regulatory Status:RUO
- Cat. No.:76741-158
Mouse Prostaglandin E Synthase/MPGES-1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Prostaglandin E Synthase/MPGES-1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 0.156 - 10 ng/ml
- Sensitivity: 0.094 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Mouse Prostaglandin E Synthase/MPGES-1 ELISA kit (A75765) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Prostaglandin E Synthase/MPGES-1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Prostaglandin E Synthase/MPGES-1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Prostaglandin E Synthase/MPGES-1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Prostaglandin E Synthase/MPGES-1 Antibody, which binds the captured Prostaglandin E Synthase/MPGES-1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Prostaglandin E Synthase/MPGES-1 captured in each well. The concentration of Prostaglandin E Synthase/MPGES-1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.