Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Thyroxine/T4 ELISA Kit

Thyroxine/T4 ELISA Kit

Thyroxine/T4 ELISA kit, 96 tests
Catalog # 76740-748
Supplier:   Antibodies.com
Supplier #:  A75876-96
Thyroxine/T4 ELISA Kit
Catalog # 76740-748
Supplier:  Antibodies.com
CAS Number:  
Thyroxine/T4 ELISA Kit
Thyroxine/T4 ELISA Kit

Thyroxine/T4 ELISA kit, 96 tests

CAS Number:  
Price
$616.68
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Thyroxine/T4 ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Thyroxine/T4 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0.703 to 45 ng/ml
  • Sensitivity: 0.422 ng/ml
  • Sample type: Serum. plasma. tissue homogenates. and other biological fluids
  • Assay precision: Intra assay: CV <8%. Inter assay: CV <10%

Thyroxine/T4 ELISA kit (A75876) employs the competitive enzyme immunoassay technique for the quantitative measurement of universal Thyroxine/T4 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Thyroxine/T4 antigen. During the incubation, Thyroxine/T4 present in the samples or standards competes with the fixed amount of immobilized Thyroxine/T4 for binding sites on the Biotinylated Anti-Thyroxine/T4 Antibody. The more Thyroxine/T4 present in a sample or standard, the less Biotinylated Anti-Thyroxine/T4 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Thyroxine/T4 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Thyroxine/T4 present in each sample or standard. The concentration of Thyroxine/T4 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Description:
    Thyroxine/T4 ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Other
  • Detection Range:
    0.703 - 45 ng/ml
  • Sample Volume:
    50 μl
  • Time to Results:
    4 h 30 min
  • Detection Method:
    Colorimetric
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76740-748
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    0.422 ng/ml
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Competitive
  • Target protein:
    Thyroxine/T4
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Shelf Life:
    Store for 6 months at 4 °C
  • Cross Reactivity:
    Thyroxine/T4 ELISA kit exhibits high specificity and excellent specificity for the detection of universal Thyroxine/T4. No significant cross-reactivity or interference between Thyroxine/T4 and analogues was observed.
  • Storage Temperature:
    4 °C

Specifications