- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Dog
- Target protein:NT-proBNP
- Description:Canine NT-proBNP ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Canine NT-proBNP ELISA Kit exhibits high specificity and excellent specificity for the detection of canine NT-proBNP. No significant cross-reactivity or interference between NT-proBNP and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:31.25 - 2000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:18.75 pg/ml
- Regulatory Status:RUO
- Cat. No.:76739-828
Canine NT-proBNP ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of canine NT-proBNP in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA kit
- Detection Range: 31.25 to 2000 pg/ml
- Sensitivity: 18.75 pg/ml
- Sample Type: Serum, plasma, tissue homogenates and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Canine NT-proBNP ELISA kit (A87000) employs the sandwich enzyme immunoassay technique for the quantitative measurement of canine NT-proBNP in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for NT-proBNP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the NT-proBNP present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-NT-proBNP Antibody, which binds the captured NT-proBNP present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of NT-proBNP captured in each well. The concentration of NT-proBNP can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.