Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Vitamin K-dependent Protein S ELISA Kit
Mouse Vitamin K-dependent Protein S ELISA Kit
Catalog # 76739-808
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse Vitamin K-dependent Protein S ELISA Kit
Catalog # 76739-808
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A77202-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Vitamin K-dependent Protein S
  • Description:
    Mouse vitamin K-dependent protein S ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Mouse Vitamin K-dependent Protein S ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Vitamin K-dependent Protein S. No significant cross-reactivity or interference between Vitamin K-dependent Protein S and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.625 - 40 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.375 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76739-808

Specifications

About this item

Mouse Vitamin K-dependent Protein S ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Vitamin K-dependent Protein S in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 0.625 to 40 ng/ml
  • Sensitivity: 0.375 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Mouse Vitamin K-dependent Protein S ELISA kit (A77202) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Vitamin K-dependent Protein S in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Vitamin K-dependent Protein S has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Vitamin K-dependent Protein S present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Vitamin K-dependent Protein S Antibody, which binds the captured Vitamin K-dependent Protein S present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Vitamin K-dependent Protein S captured in each well. The concentration of Vitamin K-dependent Protein S can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.