Order Entry
Puerto Rico
ContactUsLinkComponent
Human PKA beta (catalytic subunit) ELISA Kit
Human PKA beta (catalytic subunit) ELISA Kit
Catalog # 76738-468
Supplier:  Antibodies.com
CAS Number:  
undefined
Human PKA beta (catalytic subunit) ELISA Kit
Catalog # 76738-468
Supplier:  Antibodies.com
Supplier Number:  A75753-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    PKA beta (catalytic subunit)
  • Description:
    Human PKA beta (catalytic subunit) ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human PKA beta (catalytic subunit) ELISA kit exhibits high specificity and excellent specificity for the detection of human PKA beta (catalytic subunit). No significant cross-reactivity or interference between PKA beta (catalytic subunit) and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    78.125 - 5000 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    46.875 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76738-468

Specifications

About this item

Human PKA beta (catalytic subunit) ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PKA beta (catalytic subunit) in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 78.125 to 5000 pg/ml
  • Sensitivity: 46.875 pg/ml
  • Sample type: Serum. plasma. tissue homogenates. and other biological fluids
  • Assay precision: Intra assay: CV <8%. Inter assay: CV <10%

Human PKA beta (catalytic subunit) ELISA kit (A75753) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human PKA beta (catalytic subunit) in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PKA beta (catalytic subunit) has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PKA beta (catalytic subunit) present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PKA beta (catalytic subunit) Antibody, which binds the captured PKA beta (catalytic subunit) present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PKA beta (catalytic subunit) captured in each well. The concentration of PKA beta (catalytic subunit) can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.