Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Glycosylated Hemoglobin Subunit alpha 1/2 ELISA Kit
Rat Glycosylated Hemoglobin Subunit alpha 1/2 ELISA Kit
Catalog # 76738-228
Supplier:  Antibodies.com
CAS Number:  
undefined
Rat Glycosylated Hemoglobin Subunit alpha 1/2 ELISA Kit
Catalog # 76738-228
Supplier:  Antibodies.com
Supplier Number:  A78232-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Glycosylated Hemoglobin Subunit alpha 1/2
  • Description:
    Rat Glycosylated Hemoglobin Subunit alpha 1/2 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Rat Glycosylated Hemoglobin Subunit alpha 1/2 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Glycosylated Hemoglobin Subunit alpha 1/2. No significant cross-reactivity or interference between Glycosylated Hemoglobin Subunit alpha 1/2 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    1.563 - 100 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.938 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76738-228

Specifications

About this item

Rat Glycosylated Hemoglobin Subunit alpha 1/2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Glycosylated Hemoglobin Subunit alpha 1/2 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 1.563 to 100 ng/ml
  • Sensitivity: 0.938 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat Glycosylated Hemoglobin Subunit alpha 1/2 ELISA Kit (A78232) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Glycosylated Hemoglobin Subunit alpha 1/2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Glycosylated Hemoglobin Subunit alpha 1/2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Glycosylated Hemoglobin Subunit alpha 1/2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Glycosylated Hemoglobin Subunit alpha 1/2 Antibody, which binds the captured Glycosylated Hemoglobin Subunit alpha 1/2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Glycosylated Hemoglobin Subunit alpha 1/2 captured in each well. The concentration of Glycosylated Hemoglobin Subunit alpha 1/2 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.