- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Other
- Target protein:Procollagen Type II C-Terminal Propeptide/PIICP
- Description:Porcine procollagen type II C-terminal propeptide/PIICP ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross Reactivity:Porcine Procollagen Type II C-Terminal Propeptide/PIICP ELISA Kit exhibits high specificity and excellent specificity for the detection of porcine Procollagen Type II C-Terminal Propeptide/PIICP. No significant cross-reactivity or interference between Procollagen Type II C-Terminal Propeptide/PIICP and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.469 - 30 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.281 ng/ml
- Regulatory Status:RUO
- Cat. No.:76737-828
Porcine Procollagen Type II C-Terminal Propeptide/PIICP ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of porcine Procollagen Type II C-Terminal Propeptide/PIICP in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Porcine Procollagen Type II C-Terminal Propeptide/PIICP ELISA kit (A303668) employs the sandwich enzyme immunoassay technique for the quantitative measurement of porcine Procollagen Type II C-Terminal Propeptide/PIICP in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Procollagen Type II C-Terminal Propeptide/PIICP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Procollagen Type II C-Terminal Propeptide/PIICP present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Procollagen Type II C-Terminal Propeptide/PIICP Antibody, which binds the captured Procollagen Type II C-Terminal Propeptide/PIICP present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Procollagen Type II C-Terminal Propeptide/PIICP captured in each well. The concentration of Procollagen Type II C-Terminal Propeptide/PIICP can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.