Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Tissue Polypeptide Specific Antigen ELISA Kit
  76734-168
 :  
undefined
Human Tissue Polypeptide Specific Antigen ELISA Kit
  76734-168
 :  A303050-96
 :  

 

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Tissue Polypeptide Specific Antigen
  • Description:
    Human tissue polypeptide specific antigen ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross Reactivity:
    Human Tissue Polypeptide Specific Antigen ELISA Kit exhibits high specificity and excellent specificity for the detection of human Tissue Polypeptide Specific Antigen. No significant cross-reactivity or interference between Tissue Polypeptide Specific Antigen and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    15.625 - 1000 µIU/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    9.375 µIU/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76734-168

 

 

Human Tissue Polypeptide Specific Antigen ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Tissue Polypeptide Specific Antigen in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human Tissue Polypeptide Specific Antigen ELISA kit (A303050) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Tissue Polypeptide Specific Antigen in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Tissue Polypeptide Specific Antigen has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Tissue Polypeptide Specific Antigen present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Tissue Polypeptide Specific Antigen Antibody, which binds the captured Tissue Polypeptide Specific Antigen present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Tissue Polypeptide Specific Antigen captured in each well. The concentration of Tissue Polypeptide Specific Antigen can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.