Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse SHBG ELISA Kit
Mouse SHBG ELISA Kit
Catalog # 76732-418
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse SHBG ELISA Kit
Catalog # 76732-418
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A77320-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    SHBG
  • Description:
    Mouse SHBG ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Mouse SHBG ELISA kit exhibits high specificity and excellent specificity for the detection of mouse SHBG. No significant cross-reactivity or interference between SHBG and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    31.25 - 2000 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    50 μl
  • Sensitivity:
    18.75 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76732-418

Specifications

About this item

Mouse SHBG ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse SHBG in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 31.25 - 2000 pg/ml
  • Sensitivity: 18.75 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Mouse SHBG ELISA kit (A77320) employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse SHBG in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with SHBG antigen. During the incubation, SHBG present in the samples or standards competes with the fixed amount of immobilized SHBG for binding sites on the Biotinylated Anti-SHBG Antibody. The more SHBG present in a sample or standard, the less Biotinylated Anti-SHBG Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-SHBG Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of SHBG present in each sample or standard. The concentration of SHBG can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.