Order Entry
Puerto Rico
ContactUsLinkComponent
Rat PGC1 alpha ELISA Kit
Rat PGC1 alpha ELISA Kit
Catalog # 76732-048
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Rat PGC1 alpha ELISA Kit
Catalog # 76732-048
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A75742-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    PGC1 alpha
  • Description:
    Rat PGC1 alpha ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Rat PGC1 alpha ELISA kit exhibits high specificity and excellent specificity for the detection of rat PGC1 alpha. No significant cross-reactivity or interference between PGC1 alpha and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.094 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76732-048

Specifications

About this item

Rat PGC1 alpha ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat PGC1 alpha in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0.156 to 10 ng/ml
  • Sensitivity: 0.094 ng/ml
  • Sample type: Serum. plasma. tissue homogenates. and other biological fluids
  • Assay precision: Intra assay: CV <8%. Inter assay: CV <10%

Rat PGC1 alpha ELISA kit (A75742) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat PGC1 alpha in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PGC1 alpha has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PGC1 alpha present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PGC1 alpha Antibody, which binds the captured PGC1 alpha present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PGC1 alpha captured in each well. The concentration of PGC1 alpha can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.