- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Hepcidin 25
- Description:Human hepcidin 25 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Human Hepcidin 25 ELISA kit exhibits high specificity and excellent specificity for the detection of human Hepcidin 25. No significant cross-reactivity or interference between Hepcidin 25 and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:15.625 - 1000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:9.375 pg/ml
- Regulatory Status:RUO
- Cat. No.:76731-708
Human Hepcidin 25 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Hepcidin 25 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 15.625 - 1000 pg/ml
- Sensitivity: 9.375 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Human Hepcidin 25 ELISA kit (A74285) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Hepcidin 25 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Hepcidin 25 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Hepcidin 25 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Hepcidin 25 Antibody, which binds the captured Hepcidin 25 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Hepcidin 25 captured in each well. The concentration of Hepcidin 25 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.