- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Serpin B3/SCCA
- Description:Human serpin B3/SCCA ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Human Serpin B3/SCCA ELISA kit exhibits high specificity and excellent specificity for the detection of human Serpin B3/SCCA. No significant cross-reactivity or interference between Serpin B3/SCCA and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:78.125 - 5000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:46.875 pg/ml
- Regulatory Status:RUO
- Cat. No.:76731-198
Human Serpin B3/SCCA ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Serpin B3/SCCA in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 78.125 - 5000 pg/ml
- Sensitivity: 46.875 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Human Serpin B3/SCCA ELISA kit (A74532) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Serpin B3/SCCA in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Serpin B3/SCCA has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Serpin B3/SCCA present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Serpin B3/SCCA Antibody, which binds the captured Serpin B3/SCCA present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Serpin B3/SCCA captured in each well. The concentration of Serpin B3/SCCA can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.