Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Factor D/CFD ELISA Kit
Rat Factor D/CFD ELISA Kit
Catalog # 76730-738
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Rat Factor D/CFD ELISA Kit
Catalog # 76730-738
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A80267-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Factor D/CFD
  • Description:
    Rat factor D/CFD ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Rat Factor D/CFD ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Factor D/CFD. No significant cross-reactivity or interference between Factor D/CFD and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.625 - 40 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.375 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76730-738

Specifications

About this item

Rat Factor D/CFD ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Factor D/CFD in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 0.625 to 40 ng/ml
  • Sensitivity: 0.375 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat Factor D/CFD ELISA kit (A80267) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Factor D/CFD in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Factor D/CFD has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Factor D/CFD present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Factor D/CFD Antibody, which binds the captured Factor D/CFD present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Factor D/CFD captured in each well. The concentration of Factor D/CFD can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.