Order Entry
Puerto Rico
ContactUsLinkComponent
Human RAB27B ELISA Kit
Human RAB27B ELISA Kit
Catalog # 76730-118
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human RAB27B ELISA Kit
Catalog # 76730-118
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A247303-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    RAB27B
  • Description:
    Human RAB27B ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross Reactivity:
    Human RAB27B ELISA Kit exhibits high specificity and excellent specificity for the detection of human RAB27B. No significant cross-reactivity or interference between RAB27B and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.313 - 20 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.188 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76730-118

Specifications

About this item

Human RAB27B ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human RAB27B in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human RAB27B ELISA kit (A247303) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human RAB27B in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for RAB27B has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the RAB27B present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-RAB27B Antibody, which binds the captured RAB27B present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of RAB27B captured in each well. The concentration of RAB27B can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.