- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Phosphoribosyl Pyrophosphate Amidotransferase
- Description:Rat phosphoribosyl pyrophosphate amidotransferase ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross Reactivity:Rat Phosphoribosyl Pyrophosphate Amidotransferase ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Phosphoribosyl Pyrophosphate Amidotransferase. No significant cross-reactivity or interference between Phosphoribosyl Pyrophosphate Amidotransferase and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.156 - 10 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.094 ng/ml
- Regulatory Status:RUO
- Cat. No.:76747-280
Rat Phosphoribosyl Pyrophosphate Amidotransferase ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Phosphoribosyl Pyrophosphate Amidotransferase in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Rat Phosphoribosyl Pyrophosphate Amidotransferase ELISA kit (A303832) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Phosphoribosyl Pyrophosphate Amidotransferase in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Phosphoribosyl Pyrophosphate Amidotransferase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Phosphoribosyl Pyrophosphate Amidotransferase present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Phosphoribosyl Pyrophosphate Amidotransferase Antibody, which binds the captured Phosphoribosyl Pyrophosphate Amidotransferase present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Phosphoribosyl Pyrophosphate Amidotransferase captured in each well. The concentration of Phosphoribosyl Pyrophosphate Amidotransferase can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.