- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Estrogen Sulfotransferase 1
- Description:Rat estrogen sulfotransferase 1 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Rat Estrogen Sulfotransferase 1 ELISA kit exhibits high specificity and excellent specificity for the detection of rat Estrogen Sulfotransferase 1. No significant cross-reactivity or interference between Estrogen Sulfotransferase 1 and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.313 - 20 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.188 ng/ml
- Regulatory Status:RUO
- Cat. No.:76745-480
Rat Estrogen Sulfotransferase 1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Estrogen Sulfotransferase 1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 0.313 - 20 ng/ml
- Sensitivity: 0.188 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Rat Estrogen Sulfotransferase 1 ELISA kit (A77380) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Estrogen Sulfotransferase 1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Estrogen Sulfotransferase 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Estrogen Sulfotransferase 1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Estrogen Sulfotransferase 1 Antibody, which binds the captured Estrogen Sulfotransferase 1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Estrogen Sulfotransferase 1 captured in each well. The concentration of Estrogen Sulfotransferase 1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.