Order Entry
Puerto Rico
ContactUsLinkComponent
Human TM4SF20 ELISA Kit
Human TM4SF20 ELISA Kit
Catalog # 76741-550
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human TM4SF20 ELISA Kit
Catalog # 76741-550
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A75038-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    TM4SF20
  • Description:
    Human TM4SF20 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human TM4SF20 ELISA Kit exhibits high specificity and excellent specificity for the detection of human TM4SF20. No significant cross-reactivity or interference between TM4SF20 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.313 - 20 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.188 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76741-550

Specifications

About this item

Human TM4SF20 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TM4SF20 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 0.313 to 20 ng/ml
  • Sensitivity: 0.188 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human TM4SF20 ELISA Kit (A75038) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human TM4SF20 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for TM4SF20 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the TM4SF20 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-TM4SF20 Antibody, which binds the captured TM4SF20 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of TM4SF20 captured in each well. The concentration of TM4SF20 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.