Order Entry
Puerto Rico
ContactUsLinkComponent
Rabbit Superoxide Dismutase 1 ELISA Kit
Rabbit Superoxide Dismutase 1 ELISA Kit
Catalog # 76741-440
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Rabbit Superoxide Dismutase 1 ELISA Kit
Catalog # 76741-440
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A75851-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rabbit
  • Target protein:
    Superoxide Dismutase 1
  • Description:
    Rabbit superoxide dismutase 1 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Rabbit Superoxide Dismutase 1 ELISA Kit exhibits high specificity and excellent specificity for the detection of rabbit Superoxide Dismutase 1. No significant cross-reactivity or interference between Superoxide Dismutase 1 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.781 - 50 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.469 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76741-440

Specifications

About this item

Rabbit Superoxide Dismutase 1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rabbit Superoxide Dismutase 1 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0.781 to 50 ng/ml
  • Sensitivity: 0.469 ng/ml
  • Sample type: Serum. plasma. tissue homogenates. and other biological fluids
  • Assay precision: Intra assay: CV <8%. Inter assay: CV <10%

Rabbit Superoxide Dismutase 1 ELISA Kit (A75851) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rabbit Superoxide Dismutase 1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Superoxide Dismutase 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Superoxide Dismutase 1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Superoxide Dismutase 1 Antibody, which binds the captured Superoxide Dismutase 1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Superoxide Dismutase 1 captured in each well. The concentration of Superoxide Dismutase 1 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.