- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:PDGF Soluble R alpha
- Description:Mouse PDGF soluble R alpha ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Mouse PDGF Soluble R alpha ELISA kit exhibits high specificity and excellent specificity for the detection of mouse PDGF Soluble R alpha. No significant cross-reactivity or interference between PDGF Soluble R alpha and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:15.625 - 1000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:9.375 pg/ml
- Regulatory Status:RUO
- Cat. No.:76740-130
Mouse PDGF Soluble R alpha ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse PDGF Soluble R alpha in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 15.625 - 1000 pg/ml
- Sensitivity: 9.375 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Mouse PDGF Soluble R alpha ELISA kit (A74244) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse PDGF Soluble R alpha in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PDGF Soluble R alpha has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PDGF Soluble R alpha present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PDGF Soluble R alpha Antibody, which binds the captured PDGF Soluble R alpha present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PDGF Soluble R alpha captured in each well. The concentration of PDGF Soluble R alpha can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.