Human Epstein Barr Virus Early Antigen IgG ELISA Kit
Catalog # 76739-630
Supplier: ANTIBODIES.COM LLC
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Indirect
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Epstein Barr Virus Early Antigen IgG
- Description:Human Epstein Barr virus early antigen IgG ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Qualitative
- Shelf Life:Store for 6 months at 4 °C
- Storage Temperature:4 °C
- Regulatory Status:RUO
- Cat. No.:76739-630
Specifications
About this item
Human Epstein Barr Virus Early Antigen IgG ELISA kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro qualitative determination of human Epstein Barr Virus Early Antigen IgG in serum, plasma, tissue homogenates, and other biological fluids.