- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Competitive
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:GABA
- Description:Mouse GABA ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Mouse GABA ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse GABA. No significant cross-reactivity or interference between GABA and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:31.25 - 2000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:50 μl
- Sensitivity:18.75 pg/ml
- Regulatory Status:RUO
- Cat. No.:76739-530
Mouse GABA ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse GABA in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 31.25 to 2000 pg/ml
- Sensitivity: 18.75 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Mouse GABA ELISA Kit (A79046) employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse GABA in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with GABA antigen. During the incubation, GABA present in the samples or standards competes with the fixed amount of immobilized GABA for binding sites on the Biotinylated Anti-GABA Antibody. The more GABA present in a sample or standard, the less Biotinylated Anti-GABA Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-GABA Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of GABA present in each sample or standard. The concentration of GABA can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.