Specifications
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Bovine
- Target protein:Interferon alpha/beta Receptor 1
- Description:Bovine interferon alpha/beta receptor 1 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Bovine Interferon alpha/beta Receptor 1 ELISA kit exhibits high specificity and excellent specificity for the detection of bovine Interferon alpha/beta Receptor 1. No significant cross-reactivity or interference between Interferon alpha/beta Receptor 1 and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.313 - 20 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.188 ng/ml
- Regulatory Status:RUO
- Cat. No.:76737-320
Specifications
About this item
Bovine Interferon alpha/beta Receptor 1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of bovine Interferon alpha/beta Receptor 1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 0.313 - 20 ng/ml
- Sensitivity: 0.188 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Bovine Interferon alpha/beta Receptor 1 ELISA kit (A75111) employs the sandwich enzyme immunoassay technique for the quantitative measurement of bovine Interferon alpha/beta Receptor 1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Interferon alpha/beta Receptor 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Interferon alpha/beta Receptor 1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Interferon alpha/beta Receptor 1 Antibody, which binds the captured Interferon alpha/beta Receptor 1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Interferon alpha/beta Receptor 1 captured in each well. The concentration of Interferon alpha/beta Receptor 1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.