- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Dog
- Target protein:FSH
- Description:Canine FSH ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross Reactivity:Canine FSH ELISA kit exhibits high specificity and excellent specificity for the detection of canine FSH. No significant cross-reactivity or interference between FSH and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:1.25 - 80 mIU/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.75 mIU/ml
- Regulatory Status:RUO
- Cat. No.:76737-140
Canine FSH ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of canine FSH in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Detection range: 1.25 to 80 mIU/ml
- Sensitivity: 0.75 mIU/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Canine FSH ELISA kit (A302552) employs the sandwich enzyme immunoassay technique for the quantitative measurement of canine FSH in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for FSH has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the FSH present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-FSH antibody, which binds the captured FSH present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of FSH captured in each well. The concentration of FSH can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.