Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Complement C1q ELISA Kit
Mouse Complement C1q ELISA Kit
Catalog # 76735-230
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse Complement C1q ELISA Kit
Catalog # 76735-230
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A303547-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Complement C1q
  • Description:
    Mouse complement C1q ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross Reactivity:
    Mouse Complement C1q ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Complement C1q. No significant cross-reactivity or interference between Complement C1q and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    31.25 - 2000 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    18.75 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76735-230

Specifications

About this item

Mouse Complement C1q ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Complement C1q in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection Range: 31.25 to 2000 pg/ml
  • Sensitivity: 18.75 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%

Mouse Complement C1q ELISA kit (A303547) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Complement C1q in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Complement C1q has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Complement C1q present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-Complement C1q antibody, which binds the captured Complement C1q present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Complement C1q captured in each well. The concentration of Complement C1q can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.