Order Entry
Puerto Rico
ContactUsLinkComponent
Rat alpha Crosslaps/alpha CTx ELISA Kit
Rat alpha Crosslaps/alpha CTx ELISA Kit
Catalog # 76733-800
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Rat alpha Crosslaps/alpha CTx ELISA Kit
Catalog # 76733-800
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A78986-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    alpha Crosslaps/alpha CTx
  • Description:
    Rat alpha crosslaps/alpha CTx ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Rat alpha Crosslaps/alpha CTx ELISA Kit exhibits high specificity and excellent specificity for the detection of rat alpha Crosslaps/alpha CTx. No significant cross-reactivity or interference between alpha Crosslaps/alpha CTx and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    93.75 - 6000 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    50 μl
  • Sensitivity:
    56.25 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76733-800

Specifications

About this item

Rat alpha Crosslaps/alpha CTx ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of rat alpha Crosslaps/alpha CTx in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 93.75 to 6000 pg/ml
  • Sensitivity: 56.25 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat alpha Crosslaps/alpha CTx ELISA kit (A78986) employs the competitive enzyme immunoassay technique for the quantitative measurement of rat alpha Crosslaps/alpha CTx in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with alpha Crosslaps/alpha CTx antigen. During the incubation, alpha Crosslaps/alpha CTx present in the samples or standards competes with the fixed amount of immobilized alpha Crosslaps/alpha CTx for binding sites on the Biotinylated Anti-alpha Crosslaps/alpha CTx Antibody. The more alpha Crosslaps/alpha CTx present in a sample or standard, the less Biotinylated Anti-alpha Crosslaps/alpha CTx Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-alpha Crosslaps/alpha CTx Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of alpha Crosslaps/alpha CTx present in each sample or standard. The concentration of alpha Crosslaps/alpha CTx can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.