- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Creatine Kinase MT/CKMT1B
- Description:Rat creatine kinase MT/CKMT1B ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Rat Creatine Kinase MT/CKMT1B ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Creatine Kinase MT/CKMT1B. No significant cross-reactivity or interference between Creatine Kinase MT/CKMT1B and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:2.813 - 180 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:1.688 ng/ml
- Regulatory Status:RUO
- Cat. No.:76732-930
Rat Creatine Kinase MT/CKMT1B ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Creatine Kinase MT/CKMT1B in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 2.813 to 180 ng/ml
- Sensitivity: 1.688 ng/ml
- Sample type: Serum. plasma. tissue homogenates and other biological fluids
- Assay precision: Intra assay: CV <8%. Inter assay: CV <10%
Rat Creatine Kinase MT/CKMT1B ELISA kit (A76331) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Creatine Kinase MT/CKMT1B in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Creatine Kinase MT/CKMT1B has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Creatine Kinase MT/CKMT1B present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Creatine Kinase MT/CKMT1B Antibody, which binds the captured Creatine Kinase MT/CKMT1B present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Creatine Kinase MT/CKMT1B captured in each well. The concentration of Creatine Kinase MT/CKMT1B can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.