Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Human PAG608 ELISA Kit

Human PAG608 ELISA kit, 96 tests
Catalog # 76730-210
Supplier:   Antibodies.com
 :  A77520-96
Human PAG608 ELISA Kit
Catalog # 76730-210
Supplier:  Antibodies.com
CAS Number:  
Human PAG608 ELISA Kit
Human PAG608 ELISA Kit

Human PAG608 ELISA kit, 96 tests

CAS Number:  
Price
$677.87
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Human PAG608 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PAG608 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 0.156 - 10 ng/ml
  • Sensitivity: 0.094 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human PAG608 ELISA kit (A77520) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human PAG608 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PAG608 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PAG608 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PAG608 Antibody, which binds the captured PAG608 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PAG608 captured in each well. The concentration of PAG608 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Description:
    Human PAG608 ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Human
  • Detection Range:
    0.156 - 10 ng/ml
  • Sample Volume:
    100 μl
  • Time to Results:
    4 h 30 min
  • Detection Method:
    Colorimetric
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76730-210
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    0.094 ng/ml
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    PAG608
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Shelf Life:
    Store for 6 months at 4 °C
  • Cross Reactivity:
    Human PAG608 ELISA kit exhibits high specificity and excellent specificity for the detection of human PAG608. No significant cross-reactivity or interference between PAG608 and analogues was observed.
  • Storage Temperature:
    4 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.