Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant EPHA2 (from Insect cells), GST
Human Recombinant EPHA2 (from Insect cells), GST
Supplier:  Biortus
Certificates
undefined
Human Recombinant EPHA2 (from Insect cells), GST
Supplier:  Biortus

Specifications

  • Conjugation:
    GST
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt P29317
  • Protein/Peptide Name:
    EPHA2
  • Purity:
    90%
  • Sequence:
    R561-I976
  • Formulation:
    50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 5 mM DTT, 5% glycerol, 0.1 mM Trehalose
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Related Information

Frequently Bought Together