Human Recombinant DYRK2 (from E. coli), GST
: Frozen liquid on dry ice.
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q92630
- Protein/Peptide Name:DYRK2
- Purity:90%
- Sequence:L2-S601
- Formulation:50 mM Tris-HCl (pH 8.0), 500 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice