Human Recombinant DAPK3 (from Insect cells), GST
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt O43293
- Protein/Peptide Name:DAPK3
- Purity:90%
- Sequence:S2-R454
- Formulation:50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 5% glycerol, 1 mM DTT, 0.1 M Trehalose
- Shipping Temperature:Frozen liquid with dry ice