Human Recombinant CSNK1A1 (from Insect cells), GST
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P48729
- Protein Synonyms:CK1
- Protein/Peptide Name:CSNK1A1
- Purity:90%
- Sequence:A2-F337
- Shipping Temperature:Frozen liquid with dry ice