Human Recombinant CSNK1G3 (from Insect cells), GST
: Biortus
: Frozen liquid on dry ice.
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9Y6M4
- Protein/Peptide Name:CSNK1G3
- Purity:90%
- Sequence:E2-K447
- Shipping Temperature:Liquid. It is shipped out with dry ice
Frequently Bought Together