Human Recombinant CSNK1G1 (from Insect cells), GST
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9HCP0
- Protein/Peptide Name:CSNK1G1
- Purity:90%
- Sequence:D2-K422
- Shipping Temperature:Liquid. It is shipped out with dry ice