Human Recombinant CAMK4 (from Insect cells), GST
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q16566
- Protein/Peptide Name:CAMK4
- Purity:90%
- Sequence:L2-Y473
- Formulation:50 mM Tris-HCl (pH 7.5), 500 mM NaCl, 5% glycerol, 5 mM DTT, 0.1 M Trehalose
- Shipping Temperature:Frozen liquid with dry ice