Human Recombinant BLK (from Insect cells), GST
: Frozen liquid on dry ice.
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P51451
- Protein/Peptide Name:BLK
- Purity:90%
- Sequence:G2-P505
- Formulation:50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice