Human Recombinant AKT1 (from Insect cells), GST
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P31749
- Protein/Peptide Name:AKT1
- Purity:90%
- Sequence:S2-A480
- Formulation:50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice