Human Recombinant KCNH2 (from Mammalian cells), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:Human
- Gene ID:UniProt Q12809
- Protein/Peptide Name:KCNH2
- Purity:95%
- Sequence:M1-S1159 end, ΔP141-A350, ΔS871-R1005
- Formulation:20 mM HEPES pH 7.5, 300 mM KCl, 0.025% (w/v) LMNG, 0.0025% (w/v) CHS, 0.1 mg/ml POPE/POPC/POPG(3:1:1), 0.5 mM TCEP, 10 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together