Human Recombinant GABRB3 (from Mammalian cells), StrepII
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:StrepII
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:Human
- Gene ID:UniProt P28472
- Protein/Peptide Name:GABRB3
- Purity:80%
- Sequence:M1-F332, A447-N473 end
- Formulation:10 mM HEPES (pH 7.5), 150 mM NaCl, 0.005% (w/v) GDN, 0.0005% (w/v) CHS
- Shipping Temperature:Frozen liquid with dry ice