Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant GLRA3 (from Insect cells), StrepII
Human Recombinant GLRA3 (from Insect cells), StrepII
Supplier:  BIORTUS USA
Certificates
undefined
Human Recombinant GLRA3 (from Insect cells), StrepII
Supplier:  BIORTUS USA
Certificates

Specifications

  • Conjugation:
    StrepII
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt O75311
  • Protein Synonyms:
    Membrane protein
  • Protein/Peptide Name:
    GLRA3
  • Purity:
    95%
  • Sequence:
    M1–H460(Δ343–418::AGT)
  • Formulation:
    20 mM HEPES (pH 8.0), 150 mM NaCl, 0.005% GDN
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Related Information