Human Recombinant APLNR (from Insect cells), Flag/His
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Flag/His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P35414
- Protein Synonyms:Membrane protein
- Protein/Peptide Name:APLNR
- Purity:90%
- Sequence:M1-R330, T177N, C325L, C326M
- Formulation:25 mM HEPES pH 7.5, 500 mM NaCl, 5% glycerol, 0.01% (w/v) DDM, 0.002% (w/v) CHS
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together