Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant XIAP (from E. coli), Unconjugated
Human Recombinant XIAP (from E. coli), Unconjugated
Supplier:  Biortus
Certificates
undefined
Human Recombinant XIAP (from E. coli), Unconjugated
Supplier:  Biortus
Certificates

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    UniProt P98170
  • Protein Synonyms:
    IAP-like protein
  • Protein/Peptide Name:
    XIAP
  • Purity:
    90%
  • Sequence:
    S241-T356
  • Formulation:
    20 mM Tris-HCl (pH 7.0), 300 mM NaCl, 5% glycerol, 1 mM TCEP
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Related Information

Frequently Bought Together