Human Recombinant XIAP (from E. coli), GST
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt P98170
- Protein Synonyms:IAP-like protein
- Protein/Peptide Name:XIAP
- Purity:85%
- Sequence:Y120-P260
- Formulation:50 mM Tris-HCl (pH 7.5), 250 mM NaCl
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together