Human Recombinant TRIM54 (from E. coli), Unconjugated
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q9BYV2
- Protein Synonyms:RING finger protein 30
- Protein/Peptide Name:TRIM54
- Purity:0.85
- Sequence:N2-P358
- Formulation:50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice