Human Recombinant TRIM44 (from Insect cells), Unconjugated
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q96DX7
- Protein/Peptide Name:TRIM44
- Purity:0.85
- Sequence:A2-T344
- Formulation:50 mM Tris-HCl (pH 7.5), 200 mM NaCl, 520 glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice