Human Recombinant Gene Name: TCEB3 (from E. coli), Unconjugated
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q8NHG8
- Protein Synonyms:RING finger protein 202
- Protein/Peptide Name:Gene Name: TCEB3
- Purity:0.85
- Sequence:G2-D242
- Formulation:50 mM Tris-HCl (pH 7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice